Learn More
Ethylenediamine, 99+%
CAS: 107-15-3 | C2H8N2 | 60.10 g/mol
Supplier: Thermo Scientific Chemicals 220422500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ethylenediamine | |
| 11.0°C | |
| 117.0°C to 118.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4555 to 1.4575 | |
| MFCD00008204 | |
| 04, 230 | |
| 0.898 | |
| ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin | |
| PIICEJLVQHRZGT-UHFFFAOYSA-N | |
| ethane-1,2-diamine | |
| 3301 | |
| CHEBI:30347 | |
| 99+% | |
| Liquid, Fuming In Air |
| 107-15-3 | |
| 0.8980g/mL | |
| 38°C | |
| 99% min. (GC) | |
| C2H8N2 | |
| H2NCH2CH2NH2 | |
| 250 mL | |
| 01,372; 04,231; 13,157 | |
| 15, 3849 | |
| Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol, slightly soluble in ether, soluble in benzene | |
| NCCN | |
| 60.10 | |
| 1.6 mPa.s (20°C) | |
| 60.1 | |
| Extra Pure |
Chemical Identifiers
| 107-15-3 | |
| 60.10 | |
| PIICEJLVQHRZGT-UHFFFAOYSA-N | |
| 3301 | |
| ethane-1,2-diamine |
| C2H8N2 | |
| MFCD00008204 | |
| ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin | |
| CHEBI:30347 | |
| NCCN |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic in contact with skin.
May cause an allergic skin reaction.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (
GHS Signal Word: Danger
EINECSNumber : 203-468-6
RUO – Research Use Only