missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine, 99%

CAS: 107-15-3 | C2H8N2 | 60.10 g/mol

Supplier:  Thermo Scientific Chemicals 118420025

 View more versions of this product

Catalog No. AC118420025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethylenediamine
11.0°C
0.8980g/mL
38°C
98.5% min. (GC)
C2H8N2
H2NCH2CH2NH2
2.5 L
01,372; 04,231; 13,157
15, 3849
Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol, slightly soluble in ether, soluble in benzene
NCCN
60.10
1.6 mPa.s (20°C)
60.1
Extra Pure
107-15-3
Colorless to Yellow
117.0°C to 118.0°C
Authentic
Glass Bottle
1.4555 to 1.4575
MFCD00008204
04, 230
0.898
ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin
PIICEJLVQHRZGT-UHFFFAOYSA-N
ethane-1,2-diamine
3301
CHEBI:30347
99%
Liquid, Fuming In Air

Chemical Identifiers

107-15-3
60.10
PIICEJLVQHRZGT-UHFFFAOYSA-N
3301
ethane-1,2-diamine
C2H8N2
MFCD00008204
ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin
CHEBI:30347
NCCN
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic in contact with skin.
May cause an allergic skin reaction.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (

GHS Signal Word: Danger

EINECSNumber : 203-468-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only