missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Ethylenediamine

Catalog No. E2626625L
Encompass
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E2626625L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E2626625L Supplier Sigma Aldrich Supplier No. E262662.5L
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethylenediamine

Specifications

CAS 107-15-3
Melting Point 8.5°C
Density 0.899 g/mL at 25°C
Boiling Point 118°C
Molecular Formula C2H8N2
Linear Formula NH2CH2CH2NH2
Refractive Index n20/D 1.4565
MDL Number MFCD00008204
Quantity 2.5 L
Synonym 1,2-Diaminoethane
Molecular Weight (g/mol) 60.1 g/mol
Show More Show Less