missing translation for 'onlineSavingsMsg'
Learn More

Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%

Catalog No. p-3736443
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC409910250 25 g Plastic bottle
AC409911000 100 g Plastic bottle
AC409915000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC409910250 Supplier Thermo Scientific Chemicals Supplier No. 409910250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 67-42-5 | C14H24N2O10 | 380.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent

Chemical Identifiers

CAS 67-42-5
Molecular Formula C14H24N2O10
Molecular Weight (g/mol) 380.34
MDL Number MFCD00004291
InChI Key DEFVIWRASFVYLL-UHFFFAOYSA-N
Synonym egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn
PubChem CID 6207
ChEBI CHEBI:30740
IUPAC Name 2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
SMILES C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O

Specifications

Melting Point 240.0°C to 245.0°C
Color White
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula [-CH2OCH2CH2N(CH2CO2H)2]2
Quantity 25 g
Beilstein 04, IV, 2433
Merck Index 15, 3576
Solubility Information Solubility in water: slightly soluble
Formula Weight 380.34
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Ethylenebis(oxyethylenenitrilo)tetraacetic acid
Show More Show Less
EINECSNumber 200-651-2

RUO – Research Use Only