missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Monoisopropyl Ether 99.0+%, TCI America™
$29.83 - $41.02
Chemical Identifiers
| CAS | 109-59-1 |
|---|---|
| Molecular Formula | C5H12O2 |
| Molecular Weight (g/mol) | 104.15 |
| MDL Number | MFCD00002866 |
| InChI Key | HCGFUIQPSOCUHI-UHFFFAOYSA-N |
| Synonym | 2-isopropoxyethanol, isopropyl oxitol, isopropyl cellosolve, dowanal eipat, ethanol, 2-isopropoxy, isopropyl glycol, ethylene glycol isopropyl ether, isopropoxyethanol, ethanol, 2-1-methylethoxy, ucar ac |
| PubChem CID | 7996 |
| IUPAC Name | 2-(propan-2-yloxy)ethan-1-ol |
| SMILES | CC(C)OCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E011025ML
|
TCI America
E011025ML |
25 mL |
Each for $29.83
|
|
|||||
|
E0110500ML
|
TCI America
E0110500ML |
500 mL |
Each for $41.02
|
|
|||||
Chemical Identifiers
| 109-59-1 | |
| 104.15 | |
| HCGFUIQPSOCUHI-UHFFFAOYSA-N | |
| 7996 | |
| CC(C)OCCO |
| C5H12O2 | |
| MFCD00002866 | |
| 2-isopropoxyethanol, isopropyl oxitol, isopropyl cellosolve, dowanal eipat, ethanol, 2-isopropoxy, isopropyl glycol, ethylene glycol isopropyl ether, isopropoxyethanol, ethanol, 2-1-methylethoxy, ucar ac | |
| 2-(propan-2-yloxy)ethan-1-ol |
Specifications
| 109-59-1 | |
| 141°C | |
| MFCD00002866 | |
| 3271 | |
| HCGFUIQPSOCUHI-UHFFFAOYSA-N | |
| 2-(propan-2-yloxy)ethan-1-ol | |
| 7996 | |
| ≥99.0% (GC) | |
| Ethylene Glycol Monoisopropyl Ether |
| Colorless | |
| C5H12O2 | |
| 25 mL | |
| 2-isopropoxyethanol, isopropyl oxitol, isopropyl cellosolve, dowanal eipat, ethanol, 2-isopropoxy, isopropyl glycol, ethylene glycol isopropyl ether, isopropoxyethanol, ethanol, 2-1-methylethoxy, ucar ac | |
| CC(C)OCCO | |
| 104.15 | |
| 104.15 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0410&(2)-2424
RTECSNumber : KL5075000
TSCA : Yes