missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Monoisopropyl Ether 99.0+%, TCI America™

Catalog No. E0110500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0110500ML 500 mL
E011025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0110500ML Supplier TCI America Supplier No. E0110500ML

Chemical Identifiers

CAS 109-59-1
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00002866
InChI Key HCGFUIQPSOCUHI-UHFFFAOYSA-N
Synonym 2-isopropoxyethanol, isopropyl oxitol, isopropyl cellosolve, dowanal eipat, ethanol, 2-isopropoxy, isopropyl glycol, ethylene glycol isopropyl ether, isopropoxyethanol, ethanol, 2-1-methylethoxy, ucar ac
PubChem CID 7996
IUPAC Name 2-(propan-2-yloxy)ethan-1-ol
SMILES CC(C)OCCO

Specifications

Color Colorless
Boiling Point 141°C
Quantity 500 mL
UN Number 3271
Formula Weight 104.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Monoisopropyl Ether
EINECSNumber (2)-0410&(2)-2424
RTECSNumber KL5075000
TSCA Yes