missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Monoisobutyl Ether 98.0+%, TCI America™

Catalog No. E0301500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0301500ML 500 mL
E030125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0301500ML Supplier TCI America Supplier No. E0301500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4439-24-1
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00059601
InChI Key HHAPGMVKBLELOE-UHFFFAOYSA-N
Synonym 2-Isobutoxyethanol, Isobutyl Cellosolve
PubChem CID 521158
IUPAC Name 2-(2-methylpropoxy)ethanol
SMILES CC(C)COCCO

Specifications

Color Colorless
Boiling Point 160°C
Quantity 500 mL
UN Number 1992
Formula Weight 118.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Monoisobutyl Ether
EINECSNumber (2)-0407&(2)-2424&(7)-0097
RTECSNumber KL3850000
TSCA Yes