missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Monobutyl Ether 99.0+%, TCI America™

Catalog No. p-7118616
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B069825ML 25 mL
B0698500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B069825ML Supplier TCI America Supplier No. B069825ML
Only null left

Chemical Identifiers

CAS 111-76-2
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00002884
InChI Key POAOYUHQDCAZBD-UHFFFAOYSA-N
Synonym 2-Butoxyethanol, Butyl Cellosolve, Butyl Glycol
PubChem CID 8133
ChEBI CHEBI:63921
IUPAC Name 2-butoxyethan-1-ol
SMILES CCCCOCCO

Specifications

Boiling Point 171°C
Melting Point -75°C
Physical Form Clear Liquid at 20°C
Color Colorless
Quantity 25 mL
UN Number 2810
Formula Weight 118.18
Percent Purity ≥99.0% (GC)
Chemical Name or Material Ethylene Glycol Monobutyl Ether
EINECSNumber (2)-0407&(2)-2424&(7)-0097
RTECSNumber KJ8575000
TSCA Yes