missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Monoacetoacetate Monomethacrylate (stabilized with BHT) 95.0+%, TCI America™

Catalog No. p-7129025
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E048925G 25 g
E0489500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E048925G Supplier TCI America Supplier No. E048925G
Only null left

Chemical Identifiers

CAS 21282-97-3
Molecular Formula C10H14O5
Molecular Weight (g/mol) 214.22
MDL Number MFCD00054405
InChI Key IBDVWXAVKPRHCU-UHFFFAOYSA-N
Synonym aaem, 2-methacryloyloxy ethyl 3-oxobutanoate, 2-methacryloyloxy ethyl acetoacetate, acetoacetoxyethyl methacrylate, unii-6l3ew3hzf2, 6l3ew3hzf2, 2-acetoacetyloxy ethyl methacrylate, hema acetoacetate, 2-2-methylacryloyl oxy ethyl 3-oxobutanoate
PubChem CID 88855
IUPAC Name 2-[(2-methylprop-2-enoyl)oxy]ethyl 3-oxobutanoate
SMILES CC(=O)CC(=O)OCCOC(=O)C(C)=C

Specifications

Color Yellow
Boiling Point 100°C
Quantity 25 g
Formula Weight 214.22
Percent Purity ≥95.0% (HPLC)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Monoacetoacetate Monomethacrylate (stabilized with BHT)
EINECSNumber (2)-3579
TSCA Yes