missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Monoacetoacetate Monomethacrylate (stabilized with BHT) 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 100°C |
| Quantity | 25 g |
| Formula Weight | 214.22 |
| Percent Purity | ≥95.0% (HPLC) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethylene Glycol Monoacetoacetate Monomethacrylate (stabilized with BHT) |
Safety and Handling
| EINECSNumber | (2)-3579 |
| TSCA | Yes |