missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Mono-tert-butyl Ether 99.0+%, TCI America™

Catalog No. p-7128956
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E035425ML 25 mL
E0354500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E035425ML Supplier TCI America Supplier No. E035425ML
Only null left

Chemical Identifiers

CAS 7580-85-0
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00059004
InChI Key BDLXTDLGTWNUFM-UHFFFAOYSA-N
Synonym 2-tert-Butoxyethanol, tert-Butyl Cellosolve
PubChem CID 24232
IUPAC Name 2-(tert-butoxy)ethan-1-ol
SMILES CC(C)(C)OCCO

Specifications

Color Colorless
Boiling Point 152°C
Quantity 25 mL
UN Number 3271
Formula Weight 118.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Mono-tert-butyl Ether
EINECSNumber (2)-2424&(7)-0097
RTECSNumber KJ8750000
TSCA No