missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene glycol dimethyl ether, 99%, Extra Dry, AcroSeal(R), Thermo Scientific Chemicals
$311.55 - $311.55
Specifications
| Chemical Name or Material | Ethylene glycol dimethyl ether |
|---|---|
| CAS | 128-37-0 |
| Density | 0.8670g/mL |
| Identification | Pass Test |
| Packaging | AcroSeal™ Glass Bottle |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC610350010
|
Thermo Scientific Chemicals
610350010 |
1 L | AcroSeal™ Glass Bottle |
Each for $311.55
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ethylene glycol dimethyl ether | |
| 0.8670g/mL | |
| AcroSeal™ Glass Bottle | |
| CH3OCH2CH2OCH3 | |
| 01, 467 | |
| 01,267; 02,143; 16,229 | |
| monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether | |
| ≥99% |
| 128-37-0 | |
| Pass Test | |
| 99% | |
| Vapor Pressure: 64hPa at 20°C | |
| 50ppm max. | |
| 15, 3245 | |
| 90.12 | |
| Extra Dry |
RUO – Research Use Only