missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Ethylene glycol dimethacrylate

Catalog No. 335681500ML
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
335681500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 335681500ML Supplier Sigma Aldrich Supplier No. 335681500ML

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethylene glycol dimethylacrylate (EGDMA) is a diester formed by condensation of two equivalents of methacrylic acid and one equivalent of ethylene glycol.It is used as a monomer to prepare Hydroxyapatite/Poly methyl methacrylate composites. 1EGDMA can be used in free radical copolymer crosslinking reactions.

Specifications

CAS 97-90-5
Density 1.051 g/mL (at 25°C)
Boiling Point 98°C to 100°C (5 mmHg)
Molecular Formula C10H14O4
Refractive Index n20/D 1.454 (literature)
Linear Formula CH2=C(CH3)COOCH2CH2OCOC(CH3)=CH2
Quantity 500 mL
MDL Number MFCD00008590
Synonym 1,2-Ethanediol dimethacrylate; Ethylene dimethacrylate
Molecular Weight (g/mol) 198.22
Percent Purity 98%
Biological Activity Respiratory system
Show More Show Less