missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Dibutyl Ether 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | -69°C |
| Color | Colorless |
| Boiling Point | 202°C |
| Quantity | 25 mL |
| Formula Weight | 174.28 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethylene Glycol Dibutyl Ether |
Safety and Handling
| EINECSNumber | (2)-0420&(7)-1321 |
| RTECSNumber | KH9450000 |
| TSCA | No |