missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Diacetate 97.0+%, TCI America™
$38.51 - $59.69
Chemical Identifiers
| CAS | 111-55-7 |
|---|---|
| Molecular Formula | C6H10O4 |
| Molecular Weight (g/mol) | 146.142 |
| MDL Number | MFCD00008718 |
| InChI Key | JTXMVXSTHSMVQF-UHFFFAOYSA-N |
| Synonym | ethylene glycol diacetate, 1,2-diacetoxyethane, glycol diacetate, ethylene diacetate, ethanediol diacetate, 1,2-ethanediol, diacetate, 1,2-ethanediol diacetate, ethylene glycol, diacetate, ethylene diethanoate, aptex donor h-plus |
| PubChem CID | 8121 |
| IUPAC Name | 2-acetyloxyethyl acetate |
| SMILES | CC(=O)OCCOC(=O)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E010925ML
|
TCI America
E010925ML |
25 mL |
Each for $38.51
|
|
|||||
|
E0109500ML
|
TCI America
E0109500ML |
500 mL |
Each for $59.69
|
|
|||||
Chemical Identifiers
| 111-55-7 | |
| 146.142 | |
| JTXMVXSTHSMVQF-UHFFFAOYSA-N | |
| 8121 | |
| CC(=O)OCCOC(=O)C |
| C6H10O4 | |
| MFCD00008718 | |
| ethylene glycol diacetate, 1,2-diacetoxyethane, glycol diacetate, ethylene diacetate, ethanediol diacetate, 1,2-ethanediol, diacetate, 1,2-ethanediol diacetate, ethylene glycol, diacetate, ethylene diethanoate, aptex donor h-plus | |
| 2-acetyloxyethyl acetate |
Specifications
| 111-55-7 | |
| 190°C | |
| MFCD00008718 | |
| ethylene glycol diacetate, 1,2-diacetoxyethane, glycol diacetate, ethylene diacetate, ethanediol diacetate, 1,2-ethanediol, diacetate, 1,2-ethanediol diacetate, ethylene glycol, diacetate, ethylene diethanoate, aptex donor h-plus | |
| CC(=O)OCCOC(=O)C | |
| 146.142 | |
| 146.14 | |
| Liquid |
| Colorless | |
| C6H10O4 | |
| 25 mL | |
| JTXMVXSTHSMVQF-UHFFFAOYSA-N | |
| 2-acetyloxyethyl acetate | |
| 8121 | |
| ≥97.0% (GC) | |
| Ethylene Glycol Diacetate |
Safety and Handling
EINECSNumber : (2)-0665
RTECSNumber : KW4025000
TSCA : Yes