missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol diacetate, 97%

Catalog No. AA4191022
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4191022 100 g
AA4191036 500 g
AA419100E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4191022 Supplier Thermo Scientific Chemicals Supplier No. 04191022

CAS: 111-55-7 | C6H10O4 | 146.142 g/mol

EGDA imparts excellent flow properties in baking lacquers and enamels and where thermoplastic acrylic resins are used. It is also a good solvent for cellulosic coatings and can be used in some ink systems such as screen inks. It has found use as a perfume fixative, and has reported applications in waterborne adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-55-7
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00008718
InChI Key JTXMVXSTHSMVQF-UHFFFAOYSA-N
Synonym ethylene glycol diacetate, 1,2-diacetoxyethane, glycol diacetate, ethylene diacetate, ethanediol diacetate, 1,2-ethanediol, diacetate, 1,2-ethanediol diacetate, ethylene glycol, diacetate, ethylene diethanoate, aptex donor h-plus
PubChem CID 8121
IUPAC Name 2-acetyloxyethyl acetate
SMILES CC(=O)OCCOC(=O)C

Specifications

Melting Point -41°C
Density 1.13 g/mL
Boiling Point 186°C to 187°C
Flash Point 82°C (181°F)
Refractive Index 1.4152
Quantity 100 g
Beilstein 1762308
Merck Index 14,3799
Solubility Information Soluble in water (16.4% by weight)
Formula Weight 146.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethylene glycol diacetate
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 203-881-1
RTECSNumber KW4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only