missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Brassylate 95.0+%, TCI America™

Catalog No. E034425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E034425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E034425ML Supplier TCI America Supplier No. E034425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105-95-3
Molecular Formula C15H26O4
Molecular Weight (g/mol) 270.37
MDL Number MFCD00046977
InChI Key XRHCAGNSDHCHFJ-UHFFFAOYSA-N
Synonym ethylene brassylate, astratone, musk t, emeressence 1150, ethylene undecane dicarboxylate, ethyl brassylate, tridecanedioic acid, cyclic ethylene ester, unii-9a87hc7rod, fema no. 3543, ccris 9097
PubChem CID 61014
IUPAC Name 1,4-dioxacycloheptadecane-5,17-dione
SMILES O=C1CCCCCCCCCCCC(=O)OCCO1

Specifications

Color Colorless
Boiling Point 142°C
Quantity 25 mL
Formula Weight 270.37
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Brassylate
EINECSNumber (5)-3880
RTECSNumber YQ1927500
TSCA Yes