missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol bis-p-toluenesulfonate, 97%

Catalog No. AAB2153909
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2153909 10 g
AAB2153918 50 g
AAB2153930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2153909 Supplier Thermo Scientific Chemicals Supplier No. B2153909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6315-52-2 | C16H18O6S2 | 370.434 g/mol

Ethylene di(p-toluenesulfonate) [Ethylene glycol ditosylate] has been used in the synthesis of, cyclohexyl-fused 1,4,7-triazacyclononane, 1,2-ethylenecalix[6]arene, 1,3- and 1,4-calix[6]crown-3, 1,3- and 1,4-calix[6]crown-4, 1,4-calix[6]benzocrown-4, 1,2- and 1,3-calix[6]crown-5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylene di(p-toluenesulfonate) [Ethylene glycol ditosylate] has been used in the synthesis of, cyclohexyl-fused 1,4,7-triazacyclononane, 1,2-ethylenecalix[6]arene, 1,3- and 1,4-calix[6]crown-3, 1,3- and 1,4-calix[6]crown-4, 1,4-calix[6]benzocrown-4, 1,2- and 1,3-calix[6]crown-5.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 6315-52-2
Molecular Formula C16H18O6S2
Molecular Weight (g/mol) 370.434
MDL Number MFCD00008549
InChI Key LZIPBJBQQPZLOR-UHFFFAOYSA-N
Synonym 1,2-bis tosyloxy ethane, ethylene glycol ditosylate, ethylene ditosylate, ethane-1,2-diyl bis 4-methylbenzenesulfonate, ethylene glycol di-p-toluenesulfonate, 1,2-bis p-toluenesulfonyloxy ethane, ethylene di p-toluenesulfonate, 1,2-ethanediol, bis 4-methylbenzenesulfonate, ethylene glycol bis-p-toluenesulfonate, 2-p-tolylsulfonyloxy ethyl 4-methylbenzenesulfonate
PubChem CID 228289
IUPAC Name 2-(4-methylphenyl)sulfonyloxyethyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCOS(=O)(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 122°C to 127°C
Quantity 10 g
Solubility Information Slightly soluble in water.
Formula Weight 370.45
Percent Purity 97%
Chemical Name or Material Ethylene glycol bis-p-toluenesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only