missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol-bis-(β-Aminoethylether)-N,N,N',N'- tetraacetic acid, Molecular Biology Reagent Grade, MP Biomedicals™

Catalog No. ICN19482310
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19482310 10 g
ICN19482325 25 g
ICN19482380 100 g
ICN19482390 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19482310 Supplier MP Biomedicals Inc Supplier No. 0219482310

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 67-42-5 | C14H24N2O10 | 380.35 g/mol

Chemical Identifiers

CAS 67-42-5
Molecular Formula C14H24N2O10
Molecular Weight (g/mol) 380.35
MDL Number MFCD00004291
InChI Key DEFVIWRASFVYLL-UHFFFAOYSA-N
Synonym egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn
PubChem CID 6207
ChEBI CHEBI:30740
IUPAC Name 2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
SMILES C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O

Specifications

Color White
Quantity 10 g
Formula Weight 380.4
Percent Purity 97%
Grade Molecular Biology Reagent
Physical Form Powder
Chemical Name or Material Ethylene Glycol-bis-(β-Aminoethylether)-N,N,N',N'- tetraacetic acid
EINECSNumber 200-651-2