missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol-bis-(β-aminoethylether)-N,N,N',N'-tetraacetic acid, 99.6%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219517425

 View more versions of this product

Catalog No. ICN19517425


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethylene glycol-bis-(β-aminoethylether)-N,N,N',N'-tetraacetic acid
White
25 g
DEFVIWRASFVYLL-UHFFFAOYSA-N
2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
6207
380.4
Powder
67-42-5
C14H24N2O10
egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn
C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O
380.35
CHEBI:30740
0.996

Chemical Identifiers

67-42-5
380.35
egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn
CHEBI:30740
C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O
C14H24N2O10
DEFVIWRASFVYLL-UHFFFAOYSA-N
6207
2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions