missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N',N'-tetraacetic Acid 95.0+%, TCI America™

Supplier:  TCI America E080525G

 View more versions of this product

Catalog No. E080525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N′,N′-tetraacetic Acid [for Biochemical Rese
241°C
C14H24N2O10
25 g
DEFVIWRASFVYLL-UHFFFAOYSA-N
2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
6207
380.35
Crystalline Powder
67-42-5
White
MFCD00004291
egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn
C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O
380.35
CHEBI:30740
≥95.0% (T)

Chemical Identifiers

67-42-5
380.35
DEFVIWRASFVYLL-UHFFFAOYSA-N
6207
2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
C14H24N2O10
MFCD00004291
egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn
CHEBI:30740
C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : AH3760000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions