missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N',N'-tetraacetic Acid 96.0+%, TCI America™
$208.78 - $208.78
Chemical Identifiers
| CAS | 67-42-5 |
|---|---|
| Molecular Formula | C14H24N2O10 |
| Molecular Weight (g/mol) | 380.35 |
| MDL Number | MFCD00004291 |
| InChI Key | DEFVIWRASFVYLL-UHFFFAOYSA-N |
| Synonym | egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn |
| PubChem CID | 6207 |
| ChEBI | CHEBI:30740 |
| IUPAC Name | 2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid |
| SMILES | C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O |
Chemical Identifiers
| 67-42-5 | |
| 380.35 | |
| DEFVIWRASFVYLL-UHFFFAOYSA-N | |
| 6207 | |
| 2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid |
| C14H24N2O10 | |
| MFCD00004291 | |
| egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn | |
| CHEBI:30740 | |
| C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O |
Specifications
| 67-42-5 | |
| White | |
| MFCD00004291 | |
| egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn | |
| C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O | |
| 380.35 | |
| CHEBI:30740 | |
| ≥96.0% (T) | |
| Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N′,N′-tetraacetic Acid |
| 241°C | |
| C14H24N2O10 | |
| 25 g | |
| DEFVIWRASFVYLL-UHFFFAOYSA-N | |
| 2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid | |
| 6207 | |
| 380.35 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : AH3760000
TSCA : Yes