missing translation for 'onlineSavingsMsg'
Learn More

Ethylene diacrylate, 95%, stabilized

Catalog No. AC366970050
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC366970050 5 mL Glass bottle
AC366970250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC366970050 Supplier Thermo Scientific Chemicals Supplier No. 366970050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2274-11-5 | C8H10O4 | 170.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2274-11-5
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.17
MDL Number MFCD00008629
InChI Key KUDUQBURMYMBIJ-UHFFFAOYSA-N
SMILES C=CC(=O)OCCOC(=O)C=C

Specifications

Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 62.0°C to 65.0°C (2.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4520 to 1.4560
Quantity 5 mL
Beilstein 02, 1469
Specific Gravity 1.04
Synonym 2-hydroxy-1-naphthoic acid, 1-naphthalenecarboxylic acid, 2-hydroxy, hydroxynaphthoic acid, 1-naphthoic acid, 2-hydroxy, unii-e3h4qz94uy, e3h4qz94uy, 1-naphthalenecarboxylic acid, hydroxy, 2-hydroxynaphthalenecarboxylic acid, 2-hydroxy-1-naphthoicacid, 2-hydroxynaphtoic acid
Solubility Information Solubility in water: slightly soluble
IUPAC Name 2-(prop-2-enoyloxy)ethyl prop-2-enoate
Stabilizer 100ppm MEHQ
PubChem CID 16790
Formula Weight 170.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethylene diacrylate, 90%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lastin
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
EINECSNumber 218-886-4

RUO – Research Use Only