missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Cyanohydrin 95.0+%, TCI America™

Catalog No. p-7124894
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C128525ML 25 mL
C1285500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C128525ML Supplier TCI America Supplier No. C128525ML
Only null left

Chemical Identifiers

CAS 109-78-4
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.079
MDL Number MFCD00002826
InChI Key WSGYTJNNHPZFKR-UHFFFAOYSA-N
Synonym 3-hydroxypropionitrile, hydracrylonitrile, 2-cyanoethanol, ethylene cyanohydrin, propanenitrile, 3-hydroxy, beta-cyanoethanol, glycol cyanohydrin, methanolacetonitrile, hydroxypropanenitrile, 2-hydroxycyanoethane
PubChem CID 8011
IUPAC Name 3-hydroxypropanenitrile
SMILES C(CO)C#N

Specifications

Color Yellow
Boiling Point 221°C
Quantity 25 mL
Formula Weight 71.08
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylene Cyanohydrin
EINECSNumber (2)-3064
RTECSNumber MU5250000
TSCA Yes