missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene chlorophosphate, 95%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethylene chlorophosphate is used in the syntheses of 2-methacryloyoxyethylphophocholine, phosphatidylcholines, 2′,3′-O-16-hentriacontanyliden-adenosine-5′-phosphocholine and hexadecylphosphocholine. It is also involved the synthesis of alpha-naphthylphosphorylcholine, which is used to detect phospholipase C activity. It is an important reactant for the synthesis of a zwitterionic silane, UV-polymerizable lipids utilizing Chabrier reaction and block copolymers of poly(aliphatic ester) clickable polyphosphoester.
Solubility
Miscible with water and chloroform.
Notes
Store in cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water and reacts violently.
Ethylene chlorophosphate is used in the syntheses of 2-methacryloyoxyethylphophocholine, phosphatidylcholines, 2′,3′-O-16-hentriacontanyliden-adenosine-5′-phosphocholine and hexadecylphosphocholine. It is also involved the synthesis of alpha-naphthylphosphorylcholine, which is used to detect phospholipase C activity. It is an important reactant for the synthesis of a zwitterionic silane, UV-polymerizable lipids utilizing Chabrier reaction and block copolymers of poly(aliphatic ester) clickable polyphosphoester.
Solubility
Miscible with water and chloroform.
Notes
Store in cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water and reacts violently.
Specifications
Specifications
| Melting Point | 12°C to 14°C |
| Density | 1.56 |
| Boiling Point | 98°C to 99°C (2 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.45 |
| Quantity | 5 g |
| UN Number | UN3265 |
| Beilstein | 606582 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Miscible with water and chloroform. |
| Show More |
Safety and Handling
| Hazard Category | H314 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S. |
| EINECSNumber | 229-560-6 |
| TSCA | No |
| Recommended Storage | Store at -20°C; Store under Nitrogen |
RUO – Research Use Only