missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Ethylene carbonate
Description
Ethylene carbonate
Specifications
Specifications
| CAS | 96-49-1 |
| Melting Point | 35°C to 38°C; 36°C (exp.) |
| Density | 1.321 g/mL (at 25°C) |
| Boiling Point | 243°C to 244°C (740 mmHg) |
| Molecular Formula | C3H4O3 |
| Linear Formula | C3H4O3 |
| Quantity | 500 g |
| MDL Number | MFCD00005382 |
| Synonym | EC; 1,3-Dioxolan-2-one |
| Molecular Weight (g/mol) | 88.06 |
| Show More |