missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Bis(thioglycolate) (Purified) 97.0+%, TCI America™

Catalog No. E133325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E133325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E133325G Supplier TCI America Supplier No. E133325G
Only null left

Chemical Identifiers

CAS 123-81-9
Molecular Formula C6H10O4S2
Molecular Weight (g/mol) 210.26
MDL Number MFCD00004875
InChI Key PSYGHMBJXWRQFD-UHFFFAOYSA-N
Synonym glycol dimercaptoacetate, ethylene mercaptoacetate, gdma, ethylene glycol bisthioglycolate, ethylenebis thioglycolate, ethylene bis thioglycolate, glycol bis mercaptoacetate, ethylene bis mercaptoacetate, ethylene glycol bis mercaptoacetate, ethylene glycol bis thioglycolic ester
PubChem CID 61059
IUPAC Name 2-[(2-sulfanylacetyl)oxy]ethyl 2-sulfanylacetate
SMILES SCC(=O)OCCOC(=O)CS

Specifications

Color Yellow
Boiling Point 139°C
Quantity 25 g
Formula Weight 210.26
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethylene Bis(thioglycolate) (Purified)
RTECSNumber AI6600000
TSCA Yes