missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ethyle2-amino-4-(4-bromophenyl)-3-thiophenecarboxylate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 306934-99-6
Molecular Formula C13H12BrNO2S
Molecular Weight (g/mol) 326.208
MDL Number MFCD00435056
InChI Key SEWFWRCESBYGFS-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-4-bromophenyl thiophene-3-carboxylate, ethyl 2-amino-4-4-bromophenyl-3-thiophenecarboxylate, ethyl 2-amino-4-4-bromo-phenyl-thiophene-3-carboxylate, 2-amino-4-4-bromo-phenyl-thiophene-3-carboxylic acid ethyl ester, 2-amino-4-4-bromophenyl-thiophene-3-carboxylic acid ethyl ester, ethyl2-amino-4-4-bromophenyl-3-thiophenecarboxylate, ethyl2-amino-4-4-bromophenyl thiophene-3-carboxylate, 2-amino-4-4-bromophenyl-3-ethoxycarbonyl thiophene, ethyl 2-amino-4-4-bromophenyl-3-thiophene carboxylate
PubChem CID 727636
IUPAC Name ethyl 2-amino-4-(4-bromophenyl)thiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC=C1C2=CC=C(C=C2)Br)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC00243DA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC00243DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC00243EA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC00243EA
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

306934-99-6
326.208
SEWFWRCESBYGFS-UHFFFAOYSA-N
727636
CCOC(=O)C1=C(SC=C1C2=CC=C(C=C2)Br)N
C13H12BrNO2S
MFCD00435056
ethyl 2-amino-4-4-bromophenyl thiophene-3-carboxylate, ethyl 2-amino-4-4-bromophenyl-3-thiophenecarboxylate, ethyl 2-amino-4-4-bromo-phenyl-thiophene-3-carboxylate, 2-amino-4-4-bromo-phenyl-thiophene-3-carboxylic acid ethyl ester, 2-amino-4-4-bromophenyl-thiophene-3-carboxylic acid ethyl ester, ethyl2-amino-4-4-bromophenyl-3-thiophenecarboxylate, ethyl2-amino-4-4-bromophenyl thiophene-3-carboxylate, 2-amino-4-4-bromophenyl-3-ethoxycarbonyl thiophene, ethyl 2-amino-4-4-bromophenyl-3-thiophene carboxylate
ethyl 2-amino-4-(4-bromophenyl)thiophene-3-carboxylate

Specifications

306934-99-6
100.0
Amber Glass Bottle
MFCD00435056
ethyl 2-amino-4-4-bromophenyl thiophene-3-carboxylate, ethyl 2-amino-4-4-bromophenyl-3-thiophenecarboxylate, ethyl 2-amino-4-4-bromo-phenyl-thiophene-3-carboxylate, 2-amino-4-4-bromo-phenyl-thiophene-3-carboxylic acid ethyl ester, 2-amino-4-4-bromophenyl-thiophene-3-carboxylic acid ethyl ester, ethyl2-amino-4-4-bromophenyl-3-thiophenecarboxylate, ethyl2-amino-4-4-bromophenyl thiophene-3-carboxylate, 2-amino-4-4-bromophenyl-3-ethoxycarbonyl thiophene, ethyl 2-amino-4-4-bromophenyl-3-thiophene carboxylate
CCOC(=O)C1=C(SC=C1C2=CC=C(C=C2)Br)N
326.208
326.21
Ethyl 2-amino-4-(4-bromophenyl)-3-thiophenecarboxylate
96.5
97%
C13H12BrNO2S
1g
SEWFWRCESBYGFS-UHFFFAOYSA-N
ethyl 2-amino-4-(4-bromophenyl)thiophene-3-carboxylate
727636
97%
SDS
missing translation for 'documents'

missing translation for 'documents'