missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-(3-methoxyphenyl)acetate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB09251EA

Quantity 10g

 View more versions of this product

Catalog No. BTB09251EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

35553-92-5
194.23
XXVVNHCWPHMLEZ-UHFFFAOYSA-N
2774971
CCOC(=O)CC1=CC(=CC=C1)OC
C11H14O3
MFCD00026897
ethyl 3-methoxyphenylacetate, ethyl 2-3-methoxyphenyl acetate, ethyl 3-methoxyphenyl acetate, benzeneacetic acid, 3-methoxy-, ethyl ester, acmc-20akzh, 3-methoxy-phenyl-acetic acid ethyl ester, ethyl 2-3-methoxyphenyl ethanoate, 3-methoxyphenylacetic acid ethyl ester, 3-methoxybenzeneacetic acid ethyl ester, 2-3-methoxyphenyl acetic acid ethyl ester
ethyl 2-(3-methoxyphenyl)acetate

Specifications

Ethyl 2-(3-methoxyphenyl)acetate
96.5
97%
C11H14O3
10g
ethyl 3-methoxyphenylacetate, ethyl 2-3-methoxyphenyl acetate, ethyl 3-methoxyphenyl acetate, benzeneacetic acid, 3-methoxy-, ethyl ester, acmc-20akzh, 3-methoxy-phenyl-acetic acid ethyl ester, ethyl 2-3-methoxyphenyl ethanoate, 3-methoxyphenylacetic acid ethyl ester, 3-methoxybenzeneacetic acid ethyl ester, 2-3-methoxyphenyl acetic acid ethyl ester
CCOC(=O)CC1=CC(=CC=C1)OC
194.23
194.23
35553-92-5
100.0
Amber Glass Bottle
CH3OC6H4CH2C(O)OCH2CH3
MFCD00026897
XXVVNHCWPHMLEZ-UHFFFAOYSA-N
ethyl 2-(3-methoxyphenyl)acetate
2774971
97%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents