missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-(3-methoxyphenyl)acetate, 97%, Thermo Scientific™

Catalog No. BTB09251DA
Change view
Click to view available options
Quantity:
1g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB09251DA 1g
BTB09251EA 10g
BTB09251EE 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTB09251DA Supplier Thermo Scientific Chemicals Supplier No. BTB09251DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35553-92-5
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00026897
InChI Key XXVVNHCWPHMLEZ-UHFFFAOYSA-N
Synonym ethyl 3-methoxyphenylacetate, ethyl 2-3-methoxyphenyl acetate, ethyl 3-methoxyphenyl acetate, benzeneacetic acid, 3-methoxy-, ethyl ester, acmc-20akzh, 3-methoxy-phenyl-acetic acid ethyl ester, ethyl 2-3-methoxyphenyl ethanoate, 3-methoxyphenylacetic acid ethyl ester, 3-methoxybenzeneacetic acid ethyl ester, 2-3-methoxyphenyl acetic acid ethyl ester
PubChem CID 2774971
IUPAC Name ethyl 2-(3-methoxyphenyl)acetate
SMILES CCOC(=O)CC1=CC(=CC=C1)OC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3OC6H4CH2C(O)OCH2CH3
Quantity 1g
Formula Weight 194.23
Percent Purity 97%
Chemical Name or Material Ethyl 2-(3-methoxyphenyl)acetate