missing translation for 'onlineSavingsMsg'
Learn More

Ethylcyclopentane 99.0+%, TCI America™

Catalog No. E02115G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E02115G 5 g
E02111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E02115G Supplier TCI America Supplier No. E02115G

Chemical Identifiers

CAS 1640-89-7
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00001389
InChI Key IFTRQJLVEBNKJK-UHFFFAOYSA-N
Synonym cyclopentane, ethyl, cyclopentane,ethyl, ethylcylopentane, acmc-1bwji, ethylcyclopentane, ethyl cyclopentane, wln: l5tj a2, 4-05-00-00104 beilstein handbook reference, iftrqjlvebnkjk-uhfffaoysa
PubChem CID 15431
IUPAC Name ethylcyclopentane
SMILES CCC1CCCC1

Specifications

Color Colorless
Boiling Point 103°C
Quantity 5 g
UN Number 3295
Formula Weight 98.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylcyclopentane
RTECSNumber GY4450000
TSCA No