missing translation for 'onlineSavingsMsg'
Learn More

Ethylcyclohexane 99.0+%, TCI America™

Catalog No. E007325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E007325ML 25 mL
E0073500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E007325ML Supplier TCI America Supplier No. E007325ML

Chemical Identifiers

CAS 1678-91-7
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00001523
InChI Key IIEWJVIFRVWJOD-UHFFFAOYSA-N
Synonym cyclohexane, ethyl, ethyl cyclohexane, cyclohexylethane, ethyl-cyclohexane, acmc-1c1qu, ethylcyclohexane, dsstox_cid_30334, dsstox_gsid_51779, wln: l6tj a2, qspl 100
PubChem CID 15504
IUPAC Name ethylcyclohexane
SMILES CCC1CCCCC1

Specifications

Color Colorless
Boiling Point 132°C
Quantity 25 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylcyclohexane
EINECSNumber (3)-2231
TSCA Yes