missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ethylboronic acid pinacol ester, 99%

Catalog No. p-7237402
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Catalog No. Quantity
AAH3686803 1 g
AAH3686806 5 g
AAH3686814 25 g
3 options

Catalog No. AAH3686803

Supplier: Thermo Scientific™ H3686803

Only null left

CAS: 82954-89-0 | C8H17BO2 | 156.03 g/mol

Chemical Identifiers

CAS 82954-89-0
Molecular Formula C8H17BO2
Molecular Weight (g/mol) 156.03
MDL Number MFCD19441153
InChI Key NPUBDPDASOEIOA-UHFFFAOYSA-N
Synonym 2-Ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane;
IUPAC Name 2-ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CCB1OC(C)(C)C(C)(C)O1

Specifications

Boiling Point 79-81°C / 120mbar
Quantity 1 g
Formula Weight 156.03
Percent Purity 99%
Physical Form Liquid
ShelfLife 5 Years