missing translation for 'onlineSavingsMsg'
Learn More

Ethylbenzene 99.0+%, TCI America™

Catalog No. E0064500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0064500ML 500 mL
E006425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0064500ML Supplier TCI America Supplier No. E0064500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100-41-4
Molecular Formula C8H10
Molecular Weight (g/mol) 106.168
MDL Number MFCD00011647
InChI Key YNQLUTRBYVCPMQ-UHFFFAOYSA-N
Synonym phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
PubChem CID 7500
ChEBI CHEBI:16101
IUPAC Name ethylbenzene
SMILES CCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 136°C
Quantity 500 mL
UN Number 1175
Formula Weight 106.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylbenzene
EINECSNumber (3)-0028&(3)-0060
RTECSNumber DA0700000
TSCA Yes