missing translation for 'onlineSavingsMsg'
Learn More

Ethylbenzene, Honeywell Riedel-de Haën™

Puriss. p.a., ≥99.0% (GC)

$184.79 - $184.79

Chemical Identifiers

CAS 100-41-4
PubChem CID 7500
ChEBI CHEBI:16101
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
60-090-003
View Documents
Honeywell Inc
03080250ML
250 mL
Each for $184.79
Only null left
 
Description

Description

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications

Chemical Identifiers

100-41-4
7500
CHEBI:16101

Specifications

100-41-4
15 °C
1.494 - 1.496
7500
min. 99.0 %
Liquid
135°C to 137°C (1.013 hPa)
Glass Bottle
250 mL
CHEBI:16101
ACS Grade Plus
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'