missing translation for 'onlineSavingsMsg'
Learn More

Ethylal 99.0+%, TCI America™

Catalog No. p-7126224
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D045725ML 25 mL
D0457500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D045725ML Supplier TCI America Supplier No. D045725ML
Only null left

Chemical Identifiers

CAS 462-95-3
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00009246
InChI Key KLKFAASOGCDTDT-UHFFFAOYSA-N
Synonym diethoxymethane, ethylal, formaldehyde diethyl acetal, diethylformal, diethoxy methane, methane, diethoxy, ethoxymethoxy ethane, ethoxymethyl ethyl ether, 1,1-diethoxymethane, 3,5-dioxaheptene
PubChem CID 10024
IUPAC Name (ethoxymethoxy)ethane
SMILES CCOCOCC

Specifications

Color Colorless
Boiling Point 85°C
Quantity 25 mL
UN Number 2373
Formula Weight 104.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylal
RTECSNumber PA8500000
TSCA Yes