missing translation for 'onlineSavingsMsg'
Learn More

Ethylal 99.0+%, TCI America™

Catalog No. d045725ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
D045725ML 25 mL
D0457500ML 500 mL
2 options

Catalog No. D045725ML

Supplier: TCI America D045725ML

Only null left

Chemical Identifiers

CAS 462-95-3
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00009246
InChI Key KLKFAASOGCDTDT-UHFFFAOYSA-N
Synonym diethoxymethane, ethylal, formaldehyde diethyl acetal, diethylformal, diethoxy methane, methane, diethoxy, ethoxymethoxy ethane, ethoxymethyl ethyl ether, 1,1-diethoxymethane, 3,5-dioxaheptene
PubChem CID 10024
IUPAC Name (ethoxymethoxy)ethane
SMILES CCOCOCC

Specifications

Color Colorless
Boiling Point 85°C
Quantity 25 mL
UN Number 2373
Formula Weight 104.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylal
RTECSNumber PA8500000
TSCA Yes