missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Violet

CAS: 2390-59-2 | C31H42ClN3 | 492.15 g/mol

Supplier:  Thermo Scientific Chemicals J6395122

 View more versions of this product

Catalog No. AAJ6395122


Only null left

Description

Description

Ethyl violet has been used to stain pancreatic tissue and elastin in the detection of gram negative bacteria, and as a reagent for the extractive-spectrophotometric determination of anionic surfactants. It has been used in the extraction and spectrophotometric determination of copper. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl violet has been used to stain pancreatic tissue and elastin in the detection of gram negative bacteria, and as a reagent for the extractive-spectrophotometric determination of anionic surfactants. It has been used in the extraction and spectrophotometric determination of copper. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis.

Solubility
Soluble in water (10 mg/ml).

Notes
Dye content approx. 80%
Specifications

Specifications

Ethyl Violet
C31H42ClN3
4117566
Soluble in water (10mg/ml).
[Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC
492.15
492.15
100 g
2390-59-2
MFCD00011765
C.I. 42600; Basic Violet 4
JVICFMRAVNKDOE-UHFFFAOYSA-M
[4-[bis[4-(diethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;chloride
16955
Green
Powder

Chemical Identifiers

2390-59-2
492.15
JVICFMRAVNKDOE-UHFFFAOYSA-M
16955
[Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC
C31H42ClN3
MFCD00011765
C.I. 42600; Basic Violet 4
[4-[bis[4-(diethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 219-231-5

RTECSNumber : KH2682000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only