missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Violet 98.0+%, TCI America™

Catalog No. A51011G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A51011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A51011G Supplier TCI America Supplier No. A51011G

Sensitive spectrophotometric reagent for anionic surfactants.

Chemical Identifiers

CAS 2390-59-2 , 65121-93-9
Molecular Formula C31H42ClN3
Molecular Weight (g/mol) 492.15
MDL Number MFCD00011765
InChI Key JVICFMRAVNKDOE-UHFFFAOYSA-M
Synonym Basic Violet 4
PubChem CID 16955
IUPAC Name tris[4-(diethylamino)phenyl]methylium chloride
SMILES [Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC

Specifications

Color Green
Quantity 1 g
Physical Form Crystal/Powder
Formula Weight 560.29
Percent Purity ≥98.0% (T)
Grade Laboratory
Chemical Name or Material Ethyl Violet [Sensitive spectrophotometric reagent for anionic surfactants]
RTECSNumber KH2682000
TSCA Yes