missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Violet, 80%, Thermo Scientific™
$74.53 - $74.53
Chemical Identifiers
| CAS | 2390-59-2 |
|---|---|
| Molecular Formula | C31H42ClN3 |
| Molecular Weight (g/mol) | 492.15 |
| MDL Number | MFCD00011765 |
| InChI Key | JVICFMRAVNKDOE-UHFFFAOYSA-M |
| Synonym | Basic Violet 4, C.I. 42600 |
| PubChem CID | 16955 |
| IUPAC Name | [4-[bis[4-(diethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;chloride |
| SMILES | [Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC410490250
|
Thermo Scientific Chemicals
410490250 |
25 g |
N/A
|
|
|||||
Chemical Identifiers
| 2390-59-2 | |
| 492.15 | |
| JVICFMRAVNKDOE-UHFFFAOYSA-M | |
| 16955 | |
| [Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC |
| C31H42ClN3 | |
| MFCD00011765 | |
| Basic Violet 4, C.I. 42600 | |
| [4-[bis[4-(diethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;chloride |
Specifications
| 2390-59-2 | |
| C31H42ClN3 | |
| Basic Violet 4, C.I. 42600 | |
| JVICFMRAVNKDOE-UHFFFAOYSA-M | |
| [Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC | |
| 492.15 | |
| 593 to 599nm (c=0.1, H2O) | |
| 492.14 | |
| Glass bottle | |
| Ethyl Violet |
| 75% min. | |
| MFCD00011765 | |
| Solubility in water: slightly soluble. Other solubilities: soluble in methanol and ethanol | |
| Authentic | |
| [4-[bis[4-(diethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;chloride | |
| 16955 | |
| 4% max. | |
| 80% | |
| 25 g |
RUO – Research Use Only