missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Violet, 80%, Thermo Scientific™
Specifications
Specifications
| Dye Content | 75% min. |
| Infrared Spectrum | Authentic |
| Lambda Max | 593 to 599nm (c=0.1, H2O) |
| Loss on Drying | 4% max. |
| Packaging | Glass bottle |
| Solubility Information | Solubility in water: slightly soluble. Other solubilities: soluble in methanol and ethanol |
| Quantity | 25 g |
| Formula Weight | 492.14 |
| Percent Purity | 80% |
| Chemical Name or Material | Ethyl Violet |
RUO – Research Use Only