Learn More
Ethyl vinyl ketone, 97%, stab.
CAS: 1629-58-9 | C5H8O | 84.118 g/mol
Supplier: Thermo Scientific Chemicals L0682914
Description
Ethyl vinyl ketone is used as reagent for annulation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl vinyl ketone is used as reagent for annulation.
Solubility
Insoluble in water; soluble in most organic solvents.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications
| Ethyl vinyl ketone | |
| 1629-58-9 | |
| 103°C to 104°C | |
| Pungent | |
| 1.42 | |
| 25 g | |
| 1735857 | |
| Insoluble in water; soluble in most organic solvents. | |
| CCC(=O)C=C | |
| 84.118 | |
| 84.12 |
| stabilized | |
| 0.844 | |
| −6°C (21°F) | |
| C5H8O | |
| MFCD00009316 | |
| UN3286 | |
| 1-penten-3-one, ethyl vinyl ketone, ethylvinylketone, 1-pentene-3-one, ethylvinyl ketone, pentenone, ketone, ethyl vinyl, vinyl ethyl ketone, unii-r0053y1az7, fema no. 3382 | |
| JLIDVCMBCGBIEY-UHFFFAOYSA-N | |
| pent-1-en-3-one | |
| 15394 | |
| 97% |
Chemical Identifiers
| 1629-58-9 | |
| 84.118 | |
| JLIDVCMBCGBIEY-UHFFFAOYSA-N | |
| 15394 | |
| CCC(=O)C=C |
| C5H8O | |
| MFCD00009316 | |
| 1-penten-3-one, ethyl vinyl ketone, ethylvinylketone, 1-pentene-3-one, ethylvinyl ketone, pentenone, ketone, ethyl vinyl, vinyl ethyl ketone, unii-r0053y1az7, fema no. 3382 | |
| pent-1-en-3-one |
Safety and Handling
GHS H Statement
H225-H300-H310-H330-H314-H318
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
EINECSNumber : 216-624-3
RTECSNumber : SB3800000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only