missing translation for 'onlineSavingsMsg'
Learn More

Ethyl vinyl ketone, 97%, stab.

Catalog No. AAL0682906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0682906 5 g
AAL0682914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0682906 Supplier Thermo Scientific Chemicals Supplier No. L0682906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1629-58-9 | C5H8O | 84.118 g/mol

Ethyl vinyl ketone is used as reagent for annulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl vinyl ketone is used as reagent for annulation.

Solubility
Insoluble in water; soluble in most organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 1629-58-9
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00009316
InChI Key JLIDVCMBCGBIEY-UHFFFAOYSA-N
Synonym 1-penten-3-one, ethyl vinyl ketone, ethylvinylketone, 1-pentene-3-one, ethylvinyl ketone, pentenone, ketone, ethyl vinyl, vinyl ethyl ketone, unii-r0053y1az7, fema no. 3382
PubChem CID 15394
IUPAC Name pent-1-en-3-one
SMILES CCC(=O)C=C

Specifications

Density 0.844
Boiling Point 103°C to 104°C
Flash Point −6°C (21°F)
Odor Pungent
Refractive Index 1.42
Quantity 5 g
UN Number UN3286
Beilstein 1735857
Solubility Information Insoluble in water; soluble in most organic solvents.
Formula Weight 84.12
Percent Purity 97%
Chemical Name or Material Ethyl vinyl ketone, stabilized
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225-H300-H310-H330-H314-H318
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
EINECSNumber 216-624-3
RTECSNumber SB3800000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only