missing translation for 'onlineSavingsMsg'
Learn More

Ethyl uracil-5-carboxylate, 98%

Catalog No. AAH2650706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2650706 5 g
AAH2650703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2650706 Supplier Thermo Scientific Chemicals Supplier No. H2650706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 28485-17-8 | C7H8N2O4 | 184.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28485-17-8
Molecular Formula C7H8N2O4
Molecular Weight (g/mol) 184.15
MDL Number MFCD00057337
InChI Key MKNYHTGOVKPZMU-UHFFFAOYSA-N
Synonym 5-carbethoxyuracil, ethyl4-hydroxy-2-oxo-1,2-dihydropyrimidine-5-carboxylate, ethyl 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate, isoorotic acid ethyl ester, ethyl 2,4-dihydroxypyrimidine-5-carboxylate, ethyl uracil-5-carboxylate, ethyl 1,2,3,4-tetrahydro-2,4-dioxopyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-2,4-dioxo-, ethyl ester, ethyl 2,4-dioxo-1,3-dihydropyrimidine-5-carboxylate, pubchem13254
PubChem CID 101411
IUPAC Name ethyl 2,4-dioxo-1H-pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CNC(=O)NC1=O

Specifications

Quantity 5 g
Formula Weight 184.15
Percent Purity 98%
Chemical Name or Material Ethyl uracil-5-carboxylate
EINECSNumber 249-053-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only