missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Undecanoate 98.0+%, TCI America™
$67.38 - $388.89
Chemical Identifiers
| CAS | 627-90-7 |
|---|---|
| Molecular Formula | C13H26O2 |
| Molecular Weight (g/mol) | 214.35 |
| MDL Number | MFCD00008958 |
| InChI Key | IAFQYUQIAOWKSB-UHFFFAOYSA-N |
| Synonym | undecanoic acid, ethyl ester, ethyl undecylate, undecanoic acid ethyl ester, ethyl hendecanoate, ethyl n-undecanoate, n-undecanoic acid ethyl ester, fema no. 3492, ethyl undecanoate, acmc-1ayp4, undecanoic acid, ethylester |
| PubChem CID | 12327 |
| IUPAC Name | ethyl undecanoate |
| SMILES | CCCCCCCCCCC(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
U004925ML
|
TCI America
U004925ML |
25 mL |
Each for $67.38
|
|
|||||
|
U0049250ML
|
TCI America
U0049250ML |
250 mL |
Each for $388.89
|
|
|||||
Chemical Identifiers
| 627-90-7 | |
| 214.35 | |
| IAFQYUQIAOWKSB-UHFFFAOYSA-N | |
| 12327 | |
| CCCCCCCCCCC(=O)OCC |
| C13H26O2 | |
| MFCD00008958 | |
| undecanoic acid, ethyl ester, ethyl undecylate, undecanoic acid ethyl ester, ethyl hendecanoate, ethyl n-undecanoate, n-undecanoic acid ethyl ester, fema no. 3492, ethyl undecanoate, acmc-1ayp4, undecanoic acid, ethylester | |
| ethyl undecanoate |
Specifications
| 627-90-7 | |
| C13H26O2 | |
| 25 mL | |
| IAFQYUQIAOWKSB-UHFFFAOYSA-N | |
| ethyl undecanoate | |
| 12327 | |
| ≥98.0% (GC) |
| 140°C | |
| MFCD00008958 | |
| undecanoic acid, ethyl ester, ethyl undecylate, undecanoic acid ethyl ester, ethyl hendecanoate, ethyl n-undecanoate, n-undecanoic acid ethyl ester, fema no. 3492, ethyl undecanoate, acmc-1ayp4, undecanoic acid, ethylester | |
| CCCCCCCCCCC(=O)OCC | |
| 214.35 | |
| 214.35 | |
| Ethyl Undecanoate |
Safety and Handling
TSCA : Yes