missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Undecafluoroamyl Ketone 98.0+%, TCI America™

Catalog No. P13635G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13635G Supplier TCI America Supplier No. P13635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 383177-55-7
Molecular Formula C8H5F11O
Molecular Weight (g/mol) 326.11
MDL Number MFCD02093488
InChI Key VHTNJYGNFUMJTN-UHFFFAOYSA-N
Synonym 1H,1H,1H,2H,2H-Undecafluoro-3-octanone, Ethyl Perfluoroamyl Ketone, Ethyl Undecafluoropentyl Ketone, 4,4,5,5,6,6,7,7,8,8,8-Undecafluoro-3-octanone
PubChem CID 44630454
IUPAC Name 4,4,5,5,6,6,7,7,8,8,8-undecafluorooctan-3-one
SMILES CCC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Color Colorless
Quantity 5 g
UN Number 1224
Formula Weight 326.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Undecafluoroamyl Ketone