missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (Trimethylsilyl)acetate 93.0+%, TCI America™

Catalog No. T15845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15845G 5 g
T158425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15845G Supplier TCI America Supplier No. T15845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4071-88-9
Molecular Formula C7H16O2Si
Molecular Weight (g/mol) 160.288
MDL Number MFCD00009172
InChI Key QQFBQBDINHJDMN-UHFFFAOYSA-N
Synonym ethyl trimethylsilyl acetate, ethyl trimethylsilylacetate, ethyl 2-trimethylsilyl acetate, etsa, trimethylsilyl acetic acid ethyl ester, acetic acid, trimethylsilyl-, ethyl ester, pubchem16058, acmc-1ahie
PubChem CID 77687
IUPAC Name ethyl 2-trimethylsilylacetate
SMILES CCOC(=O)C[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 159°C
Quantity 5 g
UN Number 3272
Formula Weight 160.29
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl (Trimethylsilyl)acetate