missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Trifluoropyruvate 98.0+%, TCI America™

Catalog No. p-7136774
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14345G 5 g
T143425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14345G Supplier TCI America Supplier No. T14345G

Chemical Identifiers

CAS 13081-18-0
Molecular Formula C5H5F3O3
Molecular Weight (g/mol) 170.09
MDL Number MFCD00114935
InChI Key KJHQVUNUOIEYSV-UHFFFAOYSA-N
Synonym ethyl trifluoropyruvate, ethyltrifluoropyruvate, ethyl 3,3,3-trifluoropyruvate, trifluoropyruvic acid ethyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, ethyl ester, ethyl 2-oxo-3,3,3-trifluoropropanoate, 3,3,3-trifluoro-2-oxopropanoic acid ethyl ester, ethyl trifluorpyruvate, ethyl-trifluoropyruvate, pubchem2013
PubChem CID 2737239
IUPAC Name ethyl 3,3,3-trifluoro-2-oxopropanoate
SMILES CCOC(=O)C(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 98°C
Quantity 5 g
UN Number 1993
Formula Weight 170.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Trifluoropyruvate