missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trifluoropyruvate, 97%

CAS: 13081-18-0 | C5H5F3O3 | 170.09 g/mol

$324.82 - $324.82

Chemical Identifiers

CAS 13081-18-0
Molecular Formula C5H5F3O3
Molecular Weight (g/mol) 170.09
MDL Number MFCD00114935
InChI Key KJHQVUNUOIEYSV-UHFFFAOYSA-N
Synonym ethyl trifluoropyruvate, ethyltrifluoropyruvate, ethyl 3,3,3-trifluoropyruvate, trifluoropyruvic acid ethyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, ethyl ester, ethyl 2-oxo-3,3,3-trifluoropropanoate, 3,3,3-trifluoro-2-oxopropanoic acid ethyl ester, ethyl trifluorpyruvate, ethyl-trifluoropyruvate, pubchem2013
PubChem CID 2737239
IUPAC Name ethyl 3,3,3-trifluoro-2-oxopropanoate
SMILES CCOC(=O)C(=O)C(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2061114
View Documents
Thermo Scientific Chemicals
B2061114
25 g
Each for $324.82
Only null left
 
Description

Description

Ethyl trifluoropyruvate acts as a building block in synthetic chemistry for the synthesis of fluorine containing compounds. It is also used as a reagent for the preparation of potential biologically active compounds like highly enantioselective organocatalytic hydroxyalkylation of indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl trifluoropyruvate acts as a building block in synthetic chemistry for the synthesis of fluorine containing compounds. It is also used as a reagent for the preparation of potential biologically active compounds like highly enantioselective organocatalytic hydroxyalkylation of indoles.

Solubility
Miscible with dichloromethane.

Notes
Incompatible with strong oxidizing agents, reducing agents, acids and alkalis (bases). Store in a cool place.
Specifications

Chemical Identifiers

13081-18-0
170.09
KJHQVUNUOIEYSV-UHFFFAOYSA-N
2737239
CCOC(=O)C(=O)C(F)(F)F
C5H5F3O3
MFCD00114935
ethyl trifluoropyruvate, ethyltrifluoropyruvate, ethyl 3,3,3-trifluoropyruvate, trifluoropyruvic acid ethyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, ethyl ester, ethyl 2-oxo-3,3,3-trifluoropropanoate, 3,3,3-trifluoro-2-oxopropanoic acid ethyl ester, ethyl trifluorpyruvate, ethyl-trifluoropyruvate, pubchem2013
ethyl 3,3,3-trifluoro-2-oxopropanoate

Specifications

13081-18-0
95°C to 98°C
C5H5F3O3
MFCD00114935
UN3272
ethyl trifluoropyruvate, ethyltrifluoropyruvate, ethyl 3,3,3-trifluoropyruvate, trifluoropyruvic acid ethyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, ethyl ester, ethyl 2-oxo-3,3,3-trifluoropropanoate, 3,3,3-trifluoro-2-oxopropanoic acid ethyl ester, ethyl trifluorpyruvate, ethyl-trifluoropyruvate, pubchem2013
KJHQVUNUOIEYSV-UHFFFAOYSA-N
ethyl 3,3,3-trifluoro-2-oxopropanoate
2737239
97%
1.4
27°C (80°F)
1.3405
25 g
2087388
Miscible with dichloromethane.
CCOC(=O)C(=O)C(F)(F)F
170.09
170.09
Ethyl trifluoropyruvate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only