missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trifluoropyruvate, 97%

Catalog No. AAB2061114
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2061114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2061114 Supplier Thermo Scientific Chemicals Supplier No. B2061114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13081-18-0 | C5H5F3O3 | 170.09 g/mol

Ethyl trifluoropyruvate acts as a building block in synthetic chemistry for the synthesis of fluorine containing compounds. It is also used as a reagent for the preparation of potential biologically active compounds like highly enantioselective organocatalytic hydroxyalkylation of indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl trifluoropyruvate acts as a building block in synthetic chemistry for the synthesis of fluorine containing compounds. It is also used as a reagent for the preparation of potential biologically active compounds like highly enantioselective organocatalytic hydroxyalkylation of indoles.

Solubility
Miscible with dichloromethane.

Notes
Incompatible with strong oxidizing agents, reducing agents, acids and alkalis (bases). Store in a cool place.

Chemical Identifiers

CAS 13081-18-0
Molecular Formula C5H5F3O3
Molecular Weight (g/mol) 170.09
MDL Number MFCD00114935
InChI Key KJHQVUNUOIEYSV-UHFFFAOYSA-N
Synonym ethyl trifluoropyruvate, ethyltrifluoropyruvate, ethyl 3,3,3-trifluoropyruvate, trifluoropyruvic acid ethyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, ethyl ester, ethyl 2-oxo-3,3,3-trifluoropropanoate, 3,3,3-trifluoro-2-oxopropanoic acid ethyl ester, ethyl trifluorpyruvate, ethyl-trifluoropyruvate, pubchem2013
PubChem CID 2737239
IUPAC Name ethyl 3,3,3-trifluoro-2-oxopropanoate
SMILES CCOC(=O)C(=O)C(F)(F)F

Specifications

Density 1.4
Boiling Point 95°C to 98°C
Flash Point 27°C (80°F)
Refractive Index 1.3405
Quantity 25 g
UN Number UN3272
Beilstein 2087388
Solubility Information Miscible with dichloromethane.
Formula Weight 170.09
Percent Purity 97%
Chemical Name or Material Ethyl trifluoropyruvate
Show More Show Less
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only